Learn More
Aviva Systems Biology Corp PPP4R2 Antibody - C-terminal region (ARP42719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP4R2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PPP4R2. Alternate names: MGC131930, PP4R2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58316