Learn More
Aviva Systems Biology Corp PPP6C Antibody - N-terminal region (ARP62420_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62420_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP6C. It was validated on Western Blot. . Alternate names: FLJ92648, MGC12249, PP6, PP6CApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-677