Learn More
Aviva Systems Biology Corp PPP6R1 Antibody - N-terminal region (ARP55109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAPS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPP6R1. Alternate names: KIAA1115, MGC138185, MGC142003, PP6R1, SAP190, SAPS1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53884