Learn More
Aviva Systems Biology Corp PPP6R3 Antibody - C-terminal region (ARP62362_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62362_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP6R3. It was validated on Western Blot. . Alternate names: C11orf23, DKFZp781E17107, DKFZp781E2374, DKFZp781O2362, FLJ11058, FLJ43065, KIAA1558, MGC125711, MGC125712, PP6R3, SAP190, SAPL, SAPLa, SAPS3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-661