Learn More
MSC Vikan Pipe Exterior Brush, 20.1 in, Medium, Blue, 1 ea
Catalog No. 502867871
Encompass
This Pipe Exterior Brush effectively cleans outer surfaces of overhead pipes and ducting. Compatible with standard and telescopic handles, it's ideal for critical hygiene and food safety environments such as food manufacturing, retail, and service sectors.
  • Purpose-built for critical hygiene and food safety environments (food manufacturing, retail, service)
  • Medium bristles for wet/dry cleaning; effective for scrubbing and moving particles
  • Cleans exterior of overhead pipes, tubes, and ducts
  • Twisted bristles minimize loss
  • Compatible with extension and Euro threaded handles for extended reach and secure attachment
  • Lightweight design reduces user fatigue
Catalog No. 50-286-7871 Supplier MSC Supplier No. 81279150
This Pipe Exterior Brush effectively cleans outer surfaces of overhead pipes and ducting. Compatible with standard and telescopic handles, it's ideal for critical hygiene and food safety environments such as food manufacturing, retail, and service sectors.
  • Purpose-built for critical hygiene and food safety environments (food manufacturing, retail, service)
  • Medium bristles for wet/dry cleaning; effective for scrubbing and moving particles
  • Cleans exterior of overhead pipes, tubes, and ducts
  • Twisted bristles minimize loss
  • Compatible with extension and Euro threaded handles for extended reach and secure attachment
  • Lightweight design reduces user fatigue