Learn More
MSC Vikan Tube Brush, 2.4 In, 19.7 In, Medium, Blue
Catalog No. 502839717
Encompass
This tube brush efficiently cleans bottles, tubes, and hard-to-reach surfaces, ideal for food manufacturing and service industries. Its durable design and specialized bristles maintain hygiene.
  • Efficiently cleans bottles, tubes, and hard-to-reach areas.
  • Suitable for food manufacturing, retail, and service.
  • Hard, thick bristles dislodge stubborn soils.
  • Designed for pipe, tube, and valve cleaning.
  • Flexible stainless steel shaft reaches curves.
  • Twisted bristles minimize loss.
  • Lightweight, reducing user fatigue.
  • Easy to clean and maintain hygiene.
  • Durable for long-lasting performance.
  • Color-coded for hygienic zoning.
  • Drop-shaped hanging hole prevents liquid pooling, aids storage.
Catalog No. 50-283-9717 Supplier MSC Supplier No. 78029352
This tube brush efficiently cleans bottles, tubes, and hard-to-reach surfaces, ideal for food manufacturing and service industries. Its durable design and specialized bristles maintain hygiene.
  • Efficiently cleans bottles, tubes, and hard-to-reach areas.
  • Suitable for food manufacturing, retail, and service.
  • Hard, thick bristles dislodge stubborn soils.
  • Designed for pipe, tube, and valve cleaning.
  • Flexible stainless steel shaft reaches curves.
  • Twisted bristles minimize loss.
  • Lightweight, reducing user fatigue.
  • Easy to clean and maintain hygiene.
  • Durable for long-lasting performance.
  • Color-coded for hygienic zoning.
  • Drop-shaped hanging hole prevents liquid pooling, aids storage.