Learn More
Tisch Scientific Polypropylene PP Syringe Filters, 10.0um, 25mm, Luer-Lock/Luer Slip, Sterile, 100 per pack, SF18270
SDP
Supplier:  Tisch Scientific SF18270
Encompass
Polypropylene PP Syringe Filters, 10.0um, 25mm, Luer-Lock/Luer Slip, Sterile, 100 per pack
Catalog No. NC1713447