Learn More
Apexbio Technology LLC PPT 263717-53-9 10mg
SDP
Catalog No. 5000090847
Encompass_Preferred
PPT (263717-53-9) is a small-molecule agonist targeting estrogen receptor (ER ) It is designed to selectively activate ER thereby modulating estrogen-mediated signaling pathways PPT exerts its biological activity primarily through receptor subtype-specific agonism displaying a relative binding affinity for ER (49%) that is 410-fold higher than for estrogen receptor (ER 0 12%) Based on these pharmacological properties PPT holds research potential in studying ER -mediated cellular responses gene expression changes and investigating estrogen-related mechanisms including receptor-specific cell proliferation differentiation and physiological processes regulated through the ER signaling axis
Catalog No. 50-000-90847 Supplier Apexbio Technology LLC Supplier No. B673510
PPT (263717-53-9) is a small-molecule agonist targeting estrogen receptor (ER ) It is designed to selectively activate ER thereby modulating estrogen-mediated signaling pathways PPT exerts its biological activity primarily through receptor subtype-specific agonism displaying a relative binding affinity for ER (49%) that is 410-fold higher than for estrogen receptor (ER 0 12%) Based on these pharmacological properties PPT holds research potential in studying ER -mediated cellular responses gene expression changes and investigating estrogen-related mechanisms including receptor-specific cell proliferation differentiation and physiological processes regulated through the ER signaling axis