Learn More
Biotang Inc Accuraci* Disposable Reagent Reservoirs,100ml, 1/pk, 100/cs, sterile. Disposable reservoirs hold solutions for multichannel pipetting. Made of virgin modified polystyrene and accommodate all types of pipettors up to 12-channel widt
Catalog No. 507515029 Shop All BIOTANG Inc Products
Encompass_Preferred
Accuraci* Disposable Reagent Reservoirs,100ml, 1/pk, 100/cs, sterile. Disposable reservoirs hold solutions for multichannel pipetting. Made of virgin modified polystyrene and accommodate all types of pipettors up to 12-channel widt
Catalog No. 50-751-5029 Supplier Biotang Inc Supplier No. 90103
Accuraci* Disposable Reagent Reservoirs,100ml, 1/pk, 100/cs, sterile. Disposable reservoirs hold solutions for multichannel pipetting. Made of virgin modified polystyrene and accommodate all types of pipettors up to 12-channel widt