Learn More
Andwin Scientific PPT TP 200-1000UL CL/GRD 576PK
SDP
Supplier:  Andwin Scientific 83007384
Encompass_Preferred
The 1000 L Pipet Tips are compatible with most pipettors. This product gives the maximum sample recovery and reduces fluid retention. The tips are constructed with transparent polypropylene to allow precise viewing of samples, are nonsterile, and ar
Catalog No. NC9396277