Learn More
Medchemexpress LLC PPTN hydrochloride | 1992047-65-0 | 99.89% | 511.96 | 1 ML
SDP
Catalog No. 5000343607
Encompass_Preferred
PPTN hydrochloride is a potent, high-affinity, competitive, and highly selective P2Y14 receptor antagonist with a KB value of 434 pM. It exhibits no agonist or antagonist effect at P2Y1, P2Y2, P2Y4, P2Y6, P2Y11, P2Y12, or P2Y13 receptors. It also shows anti-inflammatory and anti-immune activity.
  • Potent, high-affinity, competitive, and highly selective P2Y14 receptor antagonist.
  • Exhibits anti-inflammatory and anti-immune activity.
  • Strong selectivity for the P2Y14-R over other nucleotide-activated P2Y receptors.
  • Inhibits UDP-glucose-promoted chemotaxis in differentiated HL-60 human promyelocytic leukemia cells with IC50s of ~1 nM (10 μM UDP-glucose) and ~4 nM (100 μM).
  • Significantly decreases ratios of p-ERK1/2 to ERK1/2 and p-p38 to p38 at 10 μM.
Catalog No. 50-003-43607 Supplier Medchemexpress LLC Supplier No. HY11032250MG
PPTN hydrochloride is a potent, high-affinity, competitive, and highly selective P2Y14 receptor antagonist with a KB value of 434 pM. It exhibits no agonist or antagonist effect at P2Y1, P2Y2, P2Y4, P2Y6, P2Y11, P2Y12, or P2Y13 receptors. It also shows anti-inflammatory and anti-immune activity.
  • Potent, high-affinity, competitive, and highly selective P2Y14 receptor antagonist.
  • Exhibits anti-inflammatory and anti-immune activity.
  • Strong selectivity for the P2Y14-R over other nucleotide-activated P2Y receptors.
  • Inhibits UDP-glucose-promoted chemotaxis in differentiated HL-60 human promyelocytic leukemia cells with IC50s of ~1 nM (10 μM UDP-glucose) and ~4 nM (100 μM).
  • Significantly decreases ratios of p-ERK1/2 to ERK1/2 and p-p38 to p38 at 10 μM.