Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PPTN hydrochloride | 1992047-65-0 | 99.9% | 511.96 | 100 MG
SDP

Catalog No. 5000343577
Encompass_Preferred

PPTN hydrochloride is a potent, high-affinity, competitive, and highly selective P2Y14 receptor antagonist with a KB value of 434 pM. It exhibits no agonist or antagonist effect at the P2Y1, P2Y2, P2Y4, P2Y6, P2Y11, P2Y12, or P2Y13 receptors. It also demonstrates anti-inflammatory and anti-immune activity.

  • Strong selectivity for the P2Y14 receptor.
  • No agonist or antagonist effect at P2Y1, P2Y2, P2Y4, P2Y6, P2Y11, P2Y12, or P2Y13 receptors.
  • Inhibits UDP-glucose-promoted chemotaxis in differentiated HL-60 human promyelocytic leukemia cells.
  • Decreases the ratios of p-ERK1/2 to ERK1/2 and p-p38 to p38.
  • Significantly decreases the expressions of NLRP3, ASC, and caspase-1 p20 in synovial tissue of rats.
  • Significantly decreases the colocalization of NLRP3 and ASC in synovial tissues of rats.

Catalog No. 50-003-43577 Supplier Medchemexpress LLC Supplier No. HY110322100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PPTN hydrochloride is a potent, high-affinity, competitive, and highly selective P2Y14 receptor antagonist with a KB value of 434 pM. It exhibits no agonist or antagonist effect at the P2Y1, P2Y2, P2Y4, P2Y6, P2Y11, P2Y12, or P2Y13 receptors. It also demonstrates anti-inflammatory and anti-immune activity.

  • Strong selectivity for the P2Y14 receptor.
  • No agonist or antagonist effect at P2Y1, P2Y2, P2Y4, P2Y6, P2Y11, P2Y12, or P2Y13 receptors.
  • Inhibits UDP-glucose-promoted chemotaxis in differentiated HL-60 human promyelocytic leukemia cells.
  • Decreases the ratios of p-ERK1/2 to ERK1/2 and p-p38 to p38.
  • Significantly decreases the expressions of NLRP3, ASC, and caspase-1 p20 in synovial tissue of rats.
  • Significantly decreases the colocalization of NLRP3 and ASC in synovial tissues of rats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.