Learn More
TARGETMOL CHEMICALS INC PPTN HYDROCHLORIDE 5MG
SDP
Catalog No. 502579419
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PPTN hydrochloride is a potent high-affinity competitive highly selective P2Y14 receptor antagonist (KB 434 pM) but no agonist or antagonist activity at P2Y1 P2Y2 P2Y4 P2Y6 P2Y11 P2Y12 or P2Y13 receptors exhibiting anti-inflammatory and anti-immune effects. purity: 99%
Catalog No. 50-257-9419 Supplier TARGETMOL CHEMICALS INC Supplier No. T635345MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PPTN hydrochloride is a potent high-affinity competitive highly selective P2Y14 receptor antagonist (KB 434 pM) but no agonist or antagonist activity at P2Y1 P2Y2 P2Y4 P2Y6 P2Y11 P2Y12 or P2Y13 receptors exhibiting anti-inflammatory and anti-immune effects. purity: 99%