Learn More
New England Biolabs, Inc. PpuMI – 2500 units
SDP
Catalog No. 50812148
Encompass
A restriction endonuclease that recognizes the sequence RG^GWC_CY.
Catalog No. 50-812-148 Supplier New England Biolabs, Inc. Supplier No. R0506L
A restriction endonuclease that recognizes the sequence RG^GWC_CY.