Learn More
New England Biolabs, Inc. PpuMI – 500 units
SDP
Catalog No. 50812149
Encompass
A restriction endonuclease that recognizes the sequence RG^GWC_CY.
Catalog No. 50-812-149 Supplier New England Biolabs, Inc. Supplier No. R0506S
A restriction endonuclease that recognizes the sequence RG^GWC_CY.