Learn More
Aviva Systems Biology Corp PPWD1 Antibody - middle region (ARP55238_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55238_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPWD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPWD1. Alternate names: KIAA0073ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61444