Learn More
Abcam PQ 401, Cell-permeable IGF1R inhibitor, 10MG
Catalog No. 502706928
Encompass_Preferred
MW 341.8 Da, Purity >99%. Cell-permeable IGF1R inhibitor (IC₅₀ = 12 μM for IGF-stimulated autophosphorylation). Shows antiproliferative effects against MCF-7 breast cancer cells. Induces renal hypertrophy in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6928 Supplier Abcam Supplier No. AB14107710MG
Add to Cart
Add to Cart
MW 341.8 Da, Purity >99%. Cell-permeable IGF1R inhibitor (IC₅₀ = 12 μM for IGF-stimulated autophosphorylation). Shows antiproliferative effects against MCF-7 breast cancer cells. Induces renal hypertrophy in vivo. .
The product is subject to the following: Abcam Restricted Use Statement