Learn More
Microsolv Technology Corporation PQ Kit, Complete. 150mm HPLC Column, Test Solutions, Accessories & Software for Performance Qualification. 1 EA.
Catalog No. 502269763
Encompass_Preferred
This complete PQ Kit™ for Self Compliance of HPLC Performance Qualification (PQ) provides everything you need to complete a full qualification of your HPLC Instruments and perform Holistic and Modular AIQ Tests. You can do Preventative Maintenance with immediate Instrument Uptime without waiting for Vendors to Verify your Instrument. Fulfill your requirements to “Maintain Instruments in a Qualified State.” See USP. You can also use this kit to Calibrate your HPLC Instruments. The HSQ Kit includes: 6 Linearity Solutions 1 Retention Time Test Solution 1 Wavelength Calibration Solution (Holmium Oxide) 1 Gradient Visualization Solution 1 Cogent HPLC Certified Test Column - 75mm 1 Software Template. Excel with Operator Determined Criteria 1 Accessories for priming pump 1 Quick Start - Instructions. The HSQ Kit Solutions will be supplied with at least six months until the expiration date. They are designed to be used promptly and often. Expiration date is two years after the da
This item is not returnable. View return policy
Catalog No. 50-226-9763 Supplier Microsolv Technology Corporation Supplier No. 4910110
This complete PQ Kit™ for Self Compliance of HPLC Performance Qualification (PQ) provides everything you need to complete a full qualification of your HPLC Instruments and perform Holistic and Modular AIQ Tests. You can do Preventative Maintenance with immediate Instrument Uptime without waiting for Vendors to Verify your Instrument. Fulfill your requirements to “Maintain Instruments in a Qualified State.” See USP. You can also use this kit to Calibrate your HPLC Instruments. The HSQ Kit includes: 6 Linearity Solutions 1 Retention Time Test Solution 1 Wavelength Calibration Solution (Holmium Oxide) 1 Gradient Visualization Solution 1 Cogent HPLC Certified Test Column - 75mm 1 Software Template. Excel with Operator Determined Criteria 1 Accessories for priming pump 1 Quick Start - Instructions. The HSQ Kit Solutions will be supplied with at least six months until the expiration date. They are designed to be used promptly and often. Expiration date is two years after the da