Learn More
Waters Corp UV Detector PQ Standard
Supplier:  Waters Corp WAT042887
Encompass_Preferred
Kit contains standards appropriate for assessment of HPLC UV systems.1 Ampule (10 mL standard solution contains):- 10.00 mg/L Acetophenone (+/- 5%) Purity > 99%- 10.00 mg/L Propiophenone (+/- 5%) Purity > 99%- 10.00 mg/L Butyrophenone (+/- 5%) Purity > 99%- 3.2 g/L Acetone (+/- 5%) Purity > 99.74%Solution in 60% HPLC Grade Methanol / 40% HPLC Grade Water, volume/volume.
Catalog No. 50-818-876