Learn More
TARGETMOL CHEMICALS INC PQ401 HYDROCHLORIDE 1968 25MG
SDP
Catalog No. 502579909
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PQ401 inhibits autophosphorylation of IGF-1R domain with IC50 of <1 uM.
Catalog No. 50-257-9909 Supplier TARGETMOL CHEMICALS INC Supplier No. T432425MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PQ401 inhibits autophosphorylation of IGF-1R domain with IC50 of <1 uM.