Learn More
Aviva Systems Biology Corp PQLC2 Antibody - middle region (ARP44918_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44918_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PQLC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PQLC2. Alternate names: FLJ20320, FLJ23906, FLJ26661, FLJ26665, RP4-657E11.5ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Pig
Catalog No. 50-014-58936