Learn More
Medchemexpress LLC HY-136660 5mg Medchemexpress, PQR626 CAS:1927857-98-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1366605MG
Encompass_Preferred
Medchemexpress, HY-136660 5mg PQR626 CAS:1927857-98-4 PQR626, a rapamycin derivative, is a potent, selective, orally active, and brain-penetrant mTOR inhibitor, with an IC50 and Ki of 5 nM and 3.6 nM, respectively. PQR626 can be can be used for the research of neurological disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1453