Learn More
Medchemexpress LLC PqsR/LasR-IN-2 | 2071705-93-4 | 98.5% | 398.89 | C21H23ClN4O2 | 5 MG
SDP
Catalog No. 5000222297
Encompass_Preferred
PqsR/LasR-IN-2 is a small-molecule research inhibitor targeting the PqsR and LasR quorum-sensing systems in Pseudomonas aeruginosa. It is provided as a purified analytical standard for in vitro studies and screening and has a reported off-target hERG inhibition (IC50 1.408 μM).
  • Potent inhibitor of PqsR and LasR in P. aeruginosa.
  • Suitable for in vitro screening and assay development.
  • Reported hERG inhibition with IC50 1.408 μM.
  • High purity (98.54%).
  • Molecular formula C21H23ClN4O2; molecular weight 398.89.
Catalog No. 50-002-22297 Supplier Medchemexpress LLC Supplier No. HY1463285MG
PqsR/LasR-IN-2 is a small-molecule research inhibitor targeting the PqsR and LasR quorum-sensing systems in Pseudomonas aeruginosa. It is provided as a purified analytical standard for in vitro studies and screening and has a reported off-target hERG inhibition (IC50 1.408 μM).
  • Potent inhibitor of PqsR and LasR in P. aeruginosa.
  • Suitable for in vitro screening and assay development.
  • Reported hERG inhibition with IC50 1.408 μM.
  • High purity (98.54%).
  • Molecular formula C21H23ClN4O2; molecular weight 398.89.