Learn More
Medchemexpress LLC PR-104 | 851627-62-8 | MFCD25976844 | 579.27 | C14H20BrN4O12PS | 10 MG
SDP
Catalog No. 5000288783
Encompass_Preferred
PR-104 is a hypoxia-activated nitrogen mustard pre-prodrug used as a DNA cross-linking agent in preclinical cancer research. It is employed in laboratory studies to evaluate hypoxia-selective cytotoxicity and DNA-damage responses in tumor models.
  • Hypoxia-activated DNA cross-linking mechanism.
  • Intended for preclinical cancer research and tumor xenograft studies.
  • Supplied in small research pack sizes suitable for analytical and in vitro work.
  • Has defined molecular formula and molecular weight for analytical characterization.
  • CAS-registered chemical for unambiguous identification and safety documentation.
Catalog No. 50-002-88783 Supplier Medchemexpress LLC Supplier No. HY1640510MG
PR-104 is a hypoxia-activated nitrogen mustard pre-prodrug used as a DNA cross-linking agent in preclinical cancer research. It is employed in laboratory studies to evaluate hypoxia-selective cytotoxicity and DNA-damage responses in tumor models.
  • Hypoxia-activated DNA cross-linking mechanism.
  • Intended for preclinical cancer research and tumor xenograft studies.
  • Supplied in small research pack sizes suitable for analytical and in vitro work.
  • Has defined molecular formula and molecular weight for analytical characterization.
  • CAS-registered chemical for unambiguous identification and safety documentation.