Learn More
Medchemexpress LLC PR-104A | 680199-06-8 | 99.0% | C14H19BrN4O9S | 1 ML
SDP
Catalog No. 5000353784
Encompass_Preferred
PR-104A (SN 27858) is the alcohol metabolite of the phosphate proagent PR-104. It functions as a hypoxia-selective DNA cross-linking and damaging agent and cytotoxin with antitumor activity. It undergoes metabolism under hypoxic conditions via the 1-electron NADPH:cytochrome P450 oxidoreductase and is utilized in research concerning relapsed/refractory T-lineage acute lymphoblastic leukemia (T-ALL).
  • Functions as a hypoxia-selective DNA cross-linking agent
  • Acts as a DNA-damaging agent and cytotoxin
  • Exhibits antitumor activity
  • Undergoes metabolism under hypoxic conditions via 1-electron NADPH:cytochrome P450 oxidoreductase
  • Utilized in research concerning relapsed/refractory T-lineage acute lymphoblastic leukemia (T-ALL)
  • For research use only
Catalog No. 50-003-53784 Supplier Medchemexpress LLC Supplier No. HY1457210MM/1ML
PR-104A (SN 27858) is the alcohol metabolite of the phosphate proagent PR-104. It functions as a hypoxia-selective DNA cross-linking and damaging agent and cytotoxin with antitumor activity. It undergoes metabolism under hypoxic conditions via the 1-electron NADPH:cytochrome P450 oxidoreductase and is utilized in research concerning relapsed/refractory T-lineage acute lymphoblastic leukemia (T-ALL).
  • Functions as a hypoxia-selective DNA cross-linking agent
  • Acts as a DNA-damaging agent and cytotoxin
  • Exhibits antitumor activity
  • Undergoes metabolism under hypoxic conditions via 1-electron NADPH:cytochrome P450 oxidoreductase
  • Utilized in research concerning relapsed/refractory T-lineage acute lymphoblastic leukemia (T-ALL)
  • For research use only