Learn More
Selleck Chemical LLC PR-619 25mg 2645-32-1
SDP
Supplier:  Selleck Chemical LLC S713025MG
Encompass_Preferred
PR-619 is a non-selective, reversible inhibitor of the deubiquitinylating enzymes (DUBs) with EC50 of 1-20 ?M in a cell-free assay. PR-619 activates autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0818370