Learn More
LifeSensors PR-619 DUB Inhibitor 5mg
SDP
Supplier:  LifeSensors SI96190005
Encompass_Preferred
PR-619 is a non-selective, reversible inhibitor of deubiquitinases/deubiquitylases (DUBs) and ubiquitin-like isopeptidases. PR-619 inhibits many deubiquitinating enzymes in the 5-20 µM range. Additionally, PR-619 is cell-permeable and treatment of cells results in the accumulation of both ubiquitin and sumo-conjugated proteins.
Catalog No. 50-103-7651