Learn More
Medchemexpress LLC Pr-924 | 1416709-79-9 | 98.9% | 618.72 | C37H38N4O5 | 25MG
SDP
Catalog No. 5000238846
Encompass_Preferred
PR-924 is a selective tripeptide epoxyketone inhibitor targeting the immunoproteasome subunit LMP-7. It covalently modifies proteasomal N-terminal threonine active sites, exhibits low-nanomolar potency (IC50 ≈ 22 nM), and has demonstrated antiproliferative and pro-apoptotic activity in multiple myeloma models. Intended for research use only.
  • Selective LMP-7 inhibition with low-nanomolar potency (IC50 ≈ 22 nM).
  • Covalent, irreversible mechanism of action for target engagement.
  • Demonstrated antiproliferative and pro-apoptotic activity in preclinical models.
  • High purity to support reproducible experimental results.
  • Available in multiple solid and solution formats for flexible dosing.
  • Supplied for research use only; not for human use.
Catalog No. 50-002-38846 Supplier Medchemexpress LLC Supplier No. HY12358725MG
PR-924 is a selective tripeptide epoxyketone inhibitor targeting the immunoproteasome subunit LMP-7. It covalently modifies proteasomal N-terminal threonine active sites, exhibits low-nanomolar potency (IC50 ≈ 22 nM), and has demonstrated antiproliferative and pro-apoptotic activity in multiple myeloma models. Intended for research use only.
  • Selective LMP-7 inhibition with low-nanomolar potency (IC50 ≈ 22 nM).
  • Covalent, irreversible mechanism of action for target engagement.
  • Demonstrated antiproliferative and pro-apoptotic activity in preclinical models.
  • High purity to support reproducible experimental results.
  • Available in multiple solid and solution formats for flexible dosing.
  • Supplied for research use only; not for human use.