Learn More
Teledyne ISCO, Inc Pr Class Dual Piston Pump With Belt Drive And Shaped Cams; 0.00 - 300.00 Ml/Min. ; 1,000 Psi
Catalog No. 502119961 Shop All Teledyne ISCO Inc. Products
Encompass
PR Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.00 - 300.00 mL/min. ; 1,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener, Pressure & Leak Sensors; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/4"""" tubing
Catalog No. 50-211-9961 Supplier Teledyne ISCO, Inc Supplier No. PR300SRX
Add to Cart
Add to Cart
Request a Quote
PR Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.00 - 300.00 mL/min. ; 1,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; with Pulse Dampener, Pressure & Leak Sensors; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" and 1/4"""" tubing