Learn More
Teledyne ISCO, Inc Pr Class Dual Piston Pump With Belt Drive And Shaped Cams; 0.0 - 100.0 Ml/Min.; 4,000 Psi
Catalog No. 502119957 Shop All Teledyne ISCO Inc. Products
Encompass
PR Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.0 - 100.0 mL/min.; 4,000 psi; PEEK Fluid Path; Automatic Piston Wash; with Pulse Dampener, Pressure & Leak Sensors; Standard Seal (Flouropolymer energizer); Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9957 Supplier Teledyne ISCO, Inc Supplier No. PR100PRX
Add to Cart
Add to Cart
Request a Quote
PR Class Dual Piston Pump with Belt Drive and Shaped Cams; 0.0 - 100.0 mL/min.; 4,000 psi; PEEK Fluid Path; Automatic Piston Wash; with Pulse Dampener, Pressure & Leak Sensors; Standard Seal (Flouropolymer energizer); Fitting package for 1/8"""" and 1/16"""" tubing