Learn More
Cell Systems Corporation 5 Vials Primary Human Coronary Artery Endothelial Cells - Same Lot (ACBRI377V5)
SDP
Catalog No. 502068282
Encompass_Preferred
Boost your research program with 5 identical vials of passage 3 Primary Human Coronary Artery Endothelial Cells while saving ~ 20% on purchase. Isolated from normal human coronary artery without positive control antibody labels. Each vial contains ~1 x 10^6 cells with 85-92% viability under ideal conditions. They are available at Passage 3 (<12 cumulative population doublings) and will initiate a Passage 4 cell culture in a 75 cm2 flask. Each vial of cells is shipped with Bac-Off (antibiotic) and CultureBoost (animal derived growth factors) at no additional cost. You can enhance consistency in your research program with reserved lots. Need vials in bulk? Contact your Fisher Scientific Sales representative for tailored quotes. Tight Deadline? Our cells are available for next day delivery after the purchase order is received. Accelerate Your Research! Try our Attachment Factor (4Z0-201, 4Z0-210) – innovative 5 seconds plating solution for all surfaces.
Catalog No. 50-206-8282 Supplier Cell Systems Corporation Supplier No. ACBRI377V5
Boost your research program with 5 identical vials of passage 3 Primary Human Coronary Artery Endothelial Cells while saving ~ 20% on purchase. Isolated from normal human coronary artery without positive control antibody labels. Each vial contains ~1 x 10^6 cells with 85-92% viability under ideal conditions. They are available at Passage 3 (<12 cumulative population doublings) and will initiate a Passage 4 cell culture in a 75 cm2 flask. Each vial of cells is shipped with Bac-Off (antibiotic) and CultureBoost (animal derived growth factors) at no additional cost. You can enhance consistency in your research program with reserved lots. Need vials in bulk? Contact your Fisher Scientific Sales representative for tailored quotes. Tight Deadline? Our cells are available for next day delivery after the purchase order is received. Accelerate Your Research! Try our Attachment Factor (4Z0-201, 4Z0-210) – innovative 5 seconds plating solution for all surfaces.