Learn More
Biogenex Laboratories PR (Insite) : PR88: 1mL
SDP
Supplier:  Biogenex Laboratories MU328UC
Encompass_Preferred
The use of monoclonal antibodies to determine progesterone receptor status increases the predictive value of immunohistochemical analysis with respect to the response of human tumors to hormonal modulation. Historically, estrogen receptor-positive/progesterone receptor-positive breast carcinoma patients have demonstrated a better response to endocrine therapy than estrogen receptor-positive/ progesterone receptor-negative patients.
Catalog No. 50-197-8884