Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PR-Set7 protein, human | 1418304-29-6 | >95.0% | 5 UG
SDP

Catalog No. 5000291564
Encompass_Preferred

PR-Set7 Protein, Human is a recombinant human histone methyltransferase supplied as a lyophilized, tag-free protein for research use. It catalyzes monomethylation of histone H4 at Lys-20 and is suitable for enzymatic and cell-based functional assays.

  • Methylates Lys-20 of histone H4.
  • Represses gene transcription and aids chromatin condensation.
  • Supports cell proliferation in functional assays.
  • Recombinant human protein expressed in E. coli and tag free.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 8.0.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.
  • Store at -20°C for long-term storage; aliquot and freeze at -80°C for extended storage.

Catalog No. 50-002-91564 Supplier Medchemexpress LLC Supplier No. HYP746135UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PR-Set7 Protein, Human is a recombinant human histone methyltransferase supplied as a lyophilized, tag-free protein for research use. It catalyzes monomethylation of histone H4 at Lys-20 and is suitable for enzymatic and cell-based functional assays.

  • Methylates Lys-20 of histone H4.
  • Represses gene transcription and aids chromatin condensation.
  • Supports cell proliferation in functional assays.
  • Recombinant human protein expressed in E. coli and tag free.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 8.0.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.
  • Store at -20°C for long-term storage; aliquot and freeze at -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.