Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PR-Set7 protein, human | >95.0% | 100 UG
SDP

Catalog No. 5000293375
Encompass_Preferred

PR-Set7 Protein, Human is a recombinant, tag-free fragment of the human PR-Set7 (KMT5A/SETD8) methyltransferase supplied as a lyophilized powder for research use. It monomethylates histone H4 at Lys20 and is provided with characterization and activity data suitable for enzymatic assays, antibody validation, and cell biology studies.

  • Recombinant human protein fragment that monomethylates histone H4 at Lys20.
  • Tag free for applications requiring unmodified protein.
  • Lyophilized powder formulation for long-term storage and convenient reconstitution.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Approx. 18 kDa molecular weight and 98 amino acids in the provided fragment.
  • Expressed in E. coli and supplied with low endotoxin (<1 EU/μg).
  • Storage recommended at -20°C; aliquot and store at -80°C for extended stability.
  • Biological activity characterized: ED50 26.51 ng/mL (cell proliferation assay).

Catalog No. 50-002-93375 Supplier Medchemexpress LLC Supplier No. HYP74613100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PR-Set7 Protein, Human is a recombinant, tag-free fragment of the human PR-Set7 (KMT5A/SETD8) methyltransferase supplied as a lyophilized powder for research use. It monomethylates histone H4 at Lys20 and is provided with characterization and activity data suitable for enzymatic assays, antibody validation, and cell biology studies.

  • Recombinant human protein fragment that monomethylates histone H4 at Lys20.
  • Tag free for applications requiring unmodified protein.
  • Lyophilized powder formulation for long-term storage and convenient reconstitution.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Approx. 18 kDa molecular weight and 98 amino acids in the provided fragment.
  • Expressed in E. coli and supplied with low endotoxin (<1 EU/μg).
  • Storage recommended at -20°C; aliquot and store at -80°C for extended stability.
  • Biological activity characterized: ED50 26.51 ng/mL (cell proliferation assay).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.