Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PR-Set7 protein, human | >95.0% | 50 UG
SDP

Catalog No. 5000291445
Encompass_Preferred

Recombinant human PR-Set7 (amino acids K195-H352) is a lyophilized, tag-free methyltransferase fragment produced in E. coli for research applications. It monomethylates histone H4 at Lys20 and is suitable for biochemical activity assays and cell-based studies.

  • Recombinant fragment K195-H352
  • Expressed in E. coli, tag free
  • Purity >95% by reducing SDS-PAGE
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 8.0
  • Molecular weight approximately 18 kDa
  • Activity ED50 = 26.51 ng/mL (specific activity 3.772x10^4 units/mg)
  • Store dry at -20°C; aliquot and freeze at -80°C for long-term storage

Catalog No. 50-002-91445 Supplier Medchemexpress LLC Supplier No. HYP7461350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PR-Set7 (amino acids K195-H352) is a lyophilized, tag-free methyltransferase fragment produced in E. coli for research applications. It monomethylates histone H4 at Lys20 and is suitable for biochemical activity assays and cell-based studies.

  • Recombinant fragment K195-H352
  • Expressed in E. coli, tag free
  • Purity >95% by reducing SDS-PAGE
  • Lyophilized from 50 mM Tris-HCl, 300 mM NaCl, pH 8.0
  • Molecular weight approximately 18 kDa
  • Activity ED50 = 26.51 ng/mL (specific activity 3.772x10^4 units/mg)
  • Store dry at -20°C; aliquot and freeze at -80°C for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.