Learn More
Enzo Life Sciences PR11 (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLPW93250100
Encompass_Preferred
Studies have demonstrated that PR39, a proline/arginine rich 39 amino acid antibacterial peptide originally derived from porcine bone marrow, exhibits a broad spectrum of biological activities, including the ability to induce angiogenesis and to limit inflammatory damage in a variety of animal models. The angiogenic effect is in part explained by the ability of PR39 to inhibit proteasome-dependent degradation of the transcription factor HIF-1a, while anti-inflammatory activity is associated with inhibition of IκBα degradation that in turn prevents activation of NFκB-dependent gene expression. The activities of PR39 reside in the N-terminal portion of the molecule encompassed by PR11. The most recent findings have demonstrated that PR39 is a non-competitive and reversible inhibitor of the proteasome function. Alternative name: N-terminal undecapeptide of PR39. Purity: ≥95% (HPLC). Solubility: Soluble in water.
Catalog No. 50-201-1679