Learn More
Kessil A Dicon Brand PR160L Mounting Module, for PR300 Shielding Box Plus
SDP
Catalog No. NC3957045
Encompass_Preferred
Kessil PR160L Mounting Module is a mechanical interface designed to securely mount Kessil PR160L-275 and PR160L-300 LED Lights into the PR300 Shielding Box Plus for a fully protected environment with efficent temperature control. This module ensures perfect lamp alignment to maintain consistent geometry and uniform irradiation during high-intensity photoreactions.
Catalog No. NC3957045 Supplier Kessil A Dicon Brand Supplier No. KSPRMM01
Kessil PR160L Mounting Module is a mechanical interface designed to securely mount Kessil PR160L-275 and PR160L-300 LED Lights into the PR300 Shielding Box Plus for a fully protected environment with efficent temperature control. This module ensures perfect lamp alignment to maintain consistent geometry and uniform irradiation during high-intensity photoreactions.