Learn More
Kessil A Dicon Brand PR300 Water Cooling System
SDP
Catalog No. NC3957038
Encompass_Preferred
Kessil PR300 Water Cooling System provides efficient cooling of reaction vessels through heat exchange with a fan or circulating ice-cold water by immersing the radiator into an ice bath. This integrated cooling assembly maintains stable reaction conditions for both parallel screening and scale-up applications. It can securely hold up to 16 x 8mL vials or 10 x 20-40mL vials to provide uniform irradiation across multiple configurations. When combined with Kessil PR300 LED Lights, it allows researchers to achieve precise process control and optimize reaction yields and product selectivity for temperature-sensitive catalysts and reagents.
Catalog No. NC3957038 Supplier Kessil A Dicon Brand Supplier No. KSPRWC01
Kessil PR300 Water Cooling System provides efficient cooling of reaction vessels through heat exchange with a fan or circulating ice-cold water by immersing the radiator into an ice bath. This integrated cooling assembly maintains stable reaction conditions for both parallel screening and scale-up applications. It can securely hold up to 16 x 8mL vials or 10 x 20-40mL vials to provide uniform irradiation across multiple configurations. When combined with Kessil PR300 LED Lights, it allows researchers to achieve precise process control and optimize reaction yields and product selectivity for temperature-sensitive catalysts and reagents.