Learn More
Enzo Life Sciences PR39 propeptide (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLPW88500100
Encompass_Preferred
A proline-arginine-rich peptide antibiotic. PR39 inhibits DNA and protein synthesis and the degradation of HIF-1α. It has been reported to bind to the α7 subunit of the 26S proteasome, blocking degradation of the NK-κB inhibitor, IκBα by the ubiquitin-proteasome pathway without affecting overall proteasome activity. More recent studies have demonstrated it to be an efficient inhibitor of all activities of the 20S proteasome. Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1648