Learn More
Medchemexpress LLC PR5-LL-CM01 | 1005307-86-7 | 98.4% | 401.51 | 200 MG
SDP
Catalog No. 5000343724
Encompass_Preferred
PR5-LL-CM01 is a potent protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 7.5 μM. It exhibits anti-tumor activities. This compound demonstrates higher efficacy in specifically inhibiting cancer cells and shows low toxicity in normal cells.
  • Potent PRMT5 inhibitor (IC50 = 7.5 μM)
  • Exhibits anti-tumor activities
  • Higher efficacy in inhibiting cancer cells with low toxicity in normal cells
  • Inhibits colony-forming ability in PANC1 and HT29 cells
  • Inhibits NF-κB activation and target gene expression in PDAC and CRC cells
  • Decreases TNFα and IL8 expression in PDAC and CRC cells
  • Displays significant anti-tumor effect in PANC1 and HT29 xenografted mice without visibly affecting body weight
Catalog No. 50-003-43724 Supplier Medchemexpress LLC Supplier No. HY10996350MG
Add to Cart
Add to Cart
PR5-LL-CM01 is a potent protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 7.5 μM. It exhibits anti-tumor activities. This compound demonstrates higher efficacy in specifically inhibiting cancer cells and shows low toxicity in normal cells.
  • Potent PRMT5 inhibitor (IC50 = 7.5 μM)
  • Exhibits anti-tumor activities
  • Higher efficacy in inhibiting cancer cells with low toxicity in normal cells
  • Inhibits colony-forming ability in PANC1 and HT29 cells
  • Inhibits NF-κB activation and target gene expression in PDAC and CRC cells
  • Decreases TNFα and IL8 expression in PDAC and CRC cells
  • Displays significant anti-tumor effect in PANC1 and HT29 xenografted mice without visibly affecting body weight