Learn More
MSC 3M 7100039246 1 Lb Bottle Clear Instant Adhesive Series PR600, 4 to 25 sec Working Time, 24 hr Full Cure Time, Bonds to Cardboard, Ceramic, Fabric, Fiberglass, Foam, Glass, Leather, Metal, Paper, Plastic, Rubber, Vinyl & Wood
Catalog No. 501305426
Encompass
3M 7100039246 1 Lb Bottle Clear Instant Adhesive Series PR600, 4 to 25 sec Working Time, 24 hr Full Cure Time, Bonds to Cardboard, Ceramic, Fabric, Fiberglass, Foam, Glass, Leather, Metal, Paper, Plastic, Rubber, Vinyl & Wood
Catalog No. 50-130-5426 Supplier MSC Supplier No. 32918070
3M 7100039246 1 Lb Bottle Clear Instant Adhesive Series PR600, 4 to 25 sec Working Time, 24 hr Full Cure Time, Bonds to Cardboard, Ceramic, Fabric, Fiberglass, Foam, Glass, Leather, Metal, Paper, Plastic, Rubber, Vinyl & Wood