Learn More
TARGETMOL CHEMICALS INC PRACINOSTAT 10MG
SDP
Catalog No. 502581065
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pracinostat (SB939) is a novel HDAC inhibitor with improved in vivo properties compared to other HDAC inhibitors currently in Clinicalal trials allowing oral dosing. Data demonstrate that Pracinostat is a potent and effective anti-tumor drug with potential as an oral therapy for a variety of human hematological and solid tumors. purity: 99%
Catalog No. 50-258-1065 Supplier TARGETMOL CHEMICALS INC Supplier No. T189010MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pracinostat (SB939) is a novel HDAC inhibitor with improved in vivo properties compared to other HDAC inhibitors currently in Clinicalal trials allowing oral dosing. Data demonstrate that Pracinostat is a potent and effective anti-tumor drug with potential as an oral therapy for a variety of human hematological and solid tumors. purity: 99%