Learn More
Apexbio Technology LLC Pracinostat (SB939) 929016-96-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063952
Encompass_Preferred
Pracinostat (SB939 CAS 929016-96-6) is an orally bioavailable inhibitor of histone deacetylases (HDAC) preferentially targeting class I II and IV HDACs with substantially lower affinity for class III enzymes By inhibiting these enzymes pracinostat promotes accumulation of acetylated histone H3 and acetylated -tubulin subsequently upregulating cell cycle regulator p21 In preclinical investigations SB939 demonstrated antiproliferative activity in various tumor cell lines including colon ovarian prostate cancers acute myeloid leukemia (AML) and B-cell lymphoma highlighting its potential utility in oncology research
Catalog No. 50-000-63952 Supplier Apexbio Technology LLC Supplier No. A40955.1
Pracinostat (SB939 CAS 929016-96-6) is an orally bioavailable inhibitor of histone deacetylases (HDAC) preferentially targeting class I II and IV HDACs with substantially lower affinity for class III enzymes By inhibiting these enzymes pracinostat promotes accumulation of acetylated histone H3 and acetylated -tubulin subsequently upregulating cell cycle regulator p21 In preclinical investigations SB939 demonstrated antiproliferative activity in various tumor cell lines including colon ovarian prostate cancers acute myeloid leukemia (AML) and B-cell lymphoma highlighting its potential utility in oncology research