Learn More
Waters Corp Book, Peptide Isolation, 72 pages, 2017
Catalog No. 502391687 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This book provides basic information about the isolation and purification of peptides. It discusses the peptide isolation workflow, method development considerations, and the use of mass-directed isolation to simplify the purification process and distinguish between target peptides and contaminants.
  • Discusses the peptide isolation workflow.
  • Covers method development considerations, including column selection, choice of mobile-phase modifier, use of temperature, and gradient optimization.
  • Addresses the use of mass-directed isolation for easier purification.
  • Authored by Jo-Ann M. Jablonski and Thomas E. Wheat.
  • Published in 2017.
  • Features 72 pages.
Catalog No. 50-239-1687 Supplier Waters Corp Supplier No. 715005429
This book provides basic information about the isolation and purification of peptides. It discusses the peptide isolation workflow, method development considerations, and the use of mass-directed isolation to simplify the purification process and distinguish between target peptides and contaminants.
  • Discusses the peptide isolation workflow.
  • Covers method development considerations, including column selection, choice of mobile-phase modifier, use of temperature, and gradient optimization.
  • Addresses the use of mass-directed isolation for easier purification.
  • Authored by Jo-Ann M. Jablonski and Thomas E. Wheat.
  • Published in 2017.
  • Features 72 pages.