Learn More
Practical Chemistry Labs

Description
- Reproducible introduction to lab equipment and techniques
- Covers principles of chemistry and research
- Use alone or with Contemporary Chemistry (S83003)
- Includes 60 labs
- Paperback, 294 pages
- Grades 9 to 12
Specifications
Specifications
| Grade | 9 to 12 |
| Format | Book |
| Product Type | Practical Chemistry Labs |
| Includes | 60 Labs |
| For Use With (Application) | Uses everyday materials for a years worth of lab sessions |
| Subject | Chemistry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.