Learn More
Medchemexpress LLC HY-119802 10mg Medchemexpress, Practolol CAS:6673-35-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11980210MG
Encompass_Preferred
Medchemexpress, HY-119802 10mg Practolol CAS:6673-35-4 Practolol is a potent and selective β1-adrenergic receptor antagonist. Practolol can be used for the research of cardiac arrhythmias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0932