Learn More
Medchemexpress LLC Pradefovir mesylate | 625095-61-6 | 99.5% | 519.90 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY112690A10MG
Encompass_Preferred
Pradefovir mesylate is a cyclodiester prodrug of PMEA, designed for efficient and specific activation by CYP3A4, an enzyme mainly located in the liver. It serves as a good substrate for liver CYP3A4 and is converted to PMEA in human liver microsomes.
  • Good substrate for liver CYP3A4
  • Converted to PMEA in human liver microsomes
  • Designed for activation through an oxidative reaction
  • Does not act as a CYP inducer in rats
Catalog No. 50-004-37769