Learn More
Medchemexpress LLC Praeruptorin A | 73069-27-9 | 50 MG
SDP
Catalog No. 5000418886
Encompass_Preferred
Praeruptorin A is a main bioactive constituent of Peucedanum praeruptorum (also known as Bai-Hua Qian Hu). Praeruptorin A exerts anti-inflammatory effects in vitro through inhibition of NF-κB activation.
  • 10 mg
  • 25 mg
  • 50 mg
  • 100 mg
Catalog No. 50-004-18886 Supplier Medchemexpress LLC Supplier No. HYN606550MG
Praeruptorin A is a main bioactive constituent of Peucedanum praeruptorum (also known as Bai-Hua Qian Hu). Praeruptorin A exerts anti-inflammatory effects in vitro through inhibition of NF-κB activation.
  • 10 mg
  • 25 mg
  • 50 mg
  • 100 mg