Learn More
Medchemexpress LLC Praeruptorin E | 78478-28-1 | 5 MG
Supplier: Medchemexpress LLC HYN60665MG
Praeruptorin E is a main bioactive constituent of Peucedanum praeruptorum, also known as Bai-Hua Qian Hu. It acts as a calcium antagonist with a pD2′ value of 5.2.
- Main bioactive constituent of Peucedanum praeruptorum.
- Functions as a calcium antagonist.
- High purity (99.18% to 99.83%).
- Soluble in DMSO (25 mg/mL).