Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Praeruptorin E | 78478-28-1 | 99.2% | 428.47 g·mol⁻¹ | C24H28O7 | 10 MG
SDP

Catalog No. 5000289741
Encompass_Preferred

Praeruptorin E is a pyranocoumarin natural product isolated from the roots of Peucedanum praeruptorum. It is used as a research reagent and is reported to have calcium channel antagonist activity; related compound Praeruptorin C has a reported pD2′ value of 5.2. The compound is supplied as a high-purity solid and is commonly offered in small milligram quantities for laboratory studies.

  • Natural pyranocoumarin isolated from Peucedanum praeruptorum.
  • Reported calcium channel antagonist activity; related compound shows pD2′ ≈ 5.2.
  • High reported purity (~99.2%), suitable for research use.
  • Available in small solid packs (for example, 10 mg) and as a 10 mM solution in DMSO.
  • Soluble in DMSO; solvent preparations available for immediate use.
  • Store at -20°C protected from light; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-89741 Supplier Medchemexpress LLC Supplier No. HYN606610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Praeruptorin E is a pyranocoumarin natural product isolated from the roots of Peucedanum praeruptorum. It is used as a research reagent and is reported to have calcium channel antagonist activity; related compound Praeruptorin C has a reported pD2′ value of 5.2. The compound is supplied as a high-purity solid and is commonly offered in small milligram quantities for laboratory studies.

  • Natural pyranocoumarin isolated from Peucedanum praeruptorum.
  • Reported calcium channel antagonist activity; related compound shows pD2′ ≈ 5.2.
  • High reported purity (~99.2%), suitable for research use.
  • Available in small solid packs (for example, 10 mg) and as a 10 mM solution in DMSO.
  • Soluble in DMSO; solvent preparations available for immediate use.
  • Store at -20°C protected from light; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.